Learn More
Biotium Alpha-Bungarotoxin, 1mg
Supplier:  Biotium 000101
Encompass_Preferred
Alpha-bungarotoxin is a potent inhibitor for the motor endplate acetylcholine receptor (AChR) (Kd = 1 nM to 1 pM). Fluorescent alpha-bungarotoxin can be used for labeling of nicotinic acetylcholine receptors at neuromuscular junctions in tissue sections. Alpha-Bungarotoxin is available conjugated to a selection of our CF dyes and other labels including biotin-XX, FITC, TAMRA, Texas Red.Properties: White solid soluble in water; Store at -20°C and protect from light; C338H529N97O105S11; MW: 7984; [11032-79-4]Texas Red is a registered trademark of Thermo Fisher Scientific
Catalog No. 50-196-4048