Learn More
Biotium Alpha-Bungarotoxin, CF568, 100ug
Supplier:  Biotium 00006100UG
Encompass_Preferred
Fluorescent conjuates of a-Bungarotoxin bind to the nicotinic acetylcholine receptor (AChR) with high affinity, and thus can be used to image the distribution of the receptor at the neuromuscular junctions. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Red fluorescent CF568 has excitation/emission at 562/583 nm.
Catalog No. 50-196-4045