Learn More
Revvity Health Sciences Inc Chromium-51 Radionuclide, 10mCi, EDTA Complex in 0.005M EDTA (pH 7.0)
Supplier:  Revvity Health Sciences Inc NEZ147010MC
Encompass
Chromium-51 Radionuclide, 10mCi, EDTA Complex in 0.005M EDTA (pH 7.0)
Catalog No. 50-281-2215
Add to Cart
Request a Quote