Learn More
Revvity Health Sciences Inc Sulfur-35 Radionuclide, 10mCi (370MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sodium Sulfate in 1mL Water
Supplier:  Revvity Health Sciences Inc NEX041H010MC
Encompass
Sulfur-35 Radionuclide, 10mCi (370MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sodium Sulfate in 1mL Water
Catalog No. 50-281-2351