Learn More
Revvity Health Sciences Inc Thyroxine, ([125I]-T4), L-[125I]-, 500µCi (18.5MBq), Specific Activity: 1080-1320µCi (40-48.8MBq)/µg
Supplier:  Revvity Health Sciences Inc NEX111H500UC
Encompass
Thyroxine, ([125I]-T4), L-[125I]-, 500µCi (18.5MBq), Specific Activity: 1080-1320µCi (40-48.8MBq)/µg
Catalog No. 50-281-2752