Learn More
Medchemexpress LLC Alphitolic acid | 19533-92-7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN28551MG
Encompass_Preferred
Alphitolic acid is an anti-inflammatory triterpene that blocks Akt-NF-κB signaling to induce apoptosis and also induces autophagy. It exhibits anti-inflammatory activity by down-regulating NO and TNF-α production, making it useful for cancer and inflammation research.
  • Anti-inflammatory triterpene
  • Induces apoptosis by blocking Akt-NF-κB signaling
  • Induces autophagy
  • Down-regulates NO and TNF-α production
  • Useful for cancer and inflammation research
Catalog No. 50-004-21425