Learn More
Medchemexpress LLC Alphitolic acid | 19533-92-7 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYN2855500UG
Encompass_Preferred
Alphitolic acid is an anti-inflammatory triterpene found in Quercus aliena. It induces apoptosis by blocking Akt-NF-κB signaling and also induces autophagy. It exhibits anti-inflammatory activity by down-regulating NO and TNF-α production and is used for cancer and inflammation research.
  • Anti-inflammatory activity
  • Induces apoptosis by blocking Akt-NF-κB signaling
  • Induces autophagy
  • Down-regulates nitric oxide and TNF-α production
  • Shows anti-proliferative activity in oral cancer cells
  • Increases p53 phosphorylation and expression
  • Decreases expression of the oncogenic E3 ligase MDM2
Catalog No. 50-004-20867