Learn More
TARGETMOL CHEMICALS INC ALPIDEM 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T266005MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alpidem (Ananxyl) is an anxiolytic drug from the imidazopyridine family. Alpidem does not produce sedative effects at normal doses and is instead used specifically for the treatment of anxiety. Alpidem has nanomolar binding affinity for both the central benzodiazepurity: 99%
Catalog No. 50-258-0686