Learn More
Cell Signaling Technology ALPP/ALPPL2 Rabbit Monoclonal Antibody, Targets ALPP/ALPPL2, Validated for WB, IHC, IF, 100 µL
SDP
Supplier:  Cell Signaling Technology 30971S
Encompass
The ALPP/ALPPL2 Rabbit Monoclonal Antibody is a highly specific detection tool for total ALPP and ALPPL2 proteins, validated for use in Western Blot, Immunofluorescence, and Immunohistochemistry applications. Engineered for high specificity, it does not cross-react with other alkaline phosphatases, making it ideal for detecting proteins often overexpressed in tumors. This antibody offers researchers a reliable and effective solution for cancer-related studies.
Catalog No. NC3307484