Learn More
Cell Signaling Technology ALPPL2 (F9B1D) Rabbit mAb #45883, 100 µl
SDP
Supplier:  Cell Signaling Technology 45883S
Encompass
ALPPL2 (F9B1D) Rabbit mAb recognizes endogenous levels of total ALPPL2 protein. This antibody does not cross-react with other ALP proteins. This antibody detects a 140 kDa protein of unknown identity in some cell lines.
Catalog No. NC3729887