Learn More
Apexbio Technology LLC Alprenolol hydrochloride 13707-88-5 100mg
SDP
Supplier:  Apexbio Technology LLC B7194100
Encompass_Preferred
Alprenolol hydrochloride (CAS 13707-88-5) is an orally active non-selective -adrenergic receptor antagonist that also exhibits antagonistic effects at 5-HT1A and 5-HT1B receptors By blocking these receptors Alprenolol hydrochloride modulates associated signaling pathways involved in the regulation of cardiovascular functions such as blood pressure control myocardial oxygen demand and cardiac rhythm This compound is utilized in pharmacological research for investigating the physiological mechanisms underlying hypertension angina pectoris and arrhythmias as well as for validating disease models related to these conditions
Catalog No. 50-000-88973