Learn More
Selleck Chemical LLC Alprenolol hydrochloride
SDP
Supplier:  Selleck Chemical LLC S58025MG
Encompass_Preferred
Alprenolol hydrochloride is the hydrochloride salt form of alprenolol which is an beta adrenergic receptorantagonist and is used as an antihypertensive anti-anginal and anti-arrhythmic agent
Catalog No. 50-000-03919