Learn More
Selleck Chemical LLC Alprostadil (PGE1) S1508-10mg
SDP
Supplier:  Selleck Chemical LLC S150810MG
Encompass_Preferred
Alprostadil (PGE1) is used as a drug in the treatment of erectile dysfunction and has vasodilatory properties It is a Prostaglandin Analog and Prostaglandin E1 Agonist
Catalog No. 50-311-1608