Learn More
Selleck Chemical LLC Alrizomadlin
SDP
Supplier:  Selleck Chemical LLC S85465MG
Encompass_Preferred
Alrizomadlin(APG-115 AA-115) is a highly potent and efficacious inhibitor of MDM2 and binds to MDM2 with IC50 of 3 8 nM It exhibits potent antiproliferative and apoptogenic activities and also significantly reduces tumor burden and prolonged survival in AML xenograft models
Catalog No. 50-000-00525