Learn More
TARGETMOL CHEMICALS INC ALS-8112 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T141951MG
Encompass_Preferred
ALS-8112 (ALS 008112) is a selective and potent inhibitor of respiratory syncytial virus (RSV) polymerase with an IC50 value of 0.02 uM. ALS-8112 exhibits antiviral activity and inhibits the RNA polymerase activity of the RSV L-P protein complex through RNA chain termination. virus infections. Purity 98.9%
Catalog No. 50-287-9588