Learn More
Cayman Chemical Alsma orIentals Alsol A 5mg
SDP
Supplier:  Cayman Chemical 340505 MG
Encompass_Preferred
A terpenoid; inhibits HBV surface antigen secretion in HBV-infected 2.2.15 cells (IC50 = 39 µM); induces cell cycle arrest at the G0/G1 phase and autophagy in MDA-MB-231 human breast cancer cells at 20 and 40 µM; reduces hepatic steatosis and increases in plasma triglyceride, total cholesterol, LDL-C, and HDL-C levels in a mouse model of high-fat diet-induced obesity at 100 mg/kg per day; decreases blood glucose and insulin levels in intraperitoneal glucose and insulin tolerance tests, respectively, in the same model
Catalog No. 50-295-0813