Learn More
Medchemexpress LLC Altanserin | 76330-71-7 | MFCD00868052 | 99.9% | 411.49 | C22H22FN3O2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY119156100MG
Encompass_Preferred
Altanserin is a selective 5-HT2A (serotonin 2A) receptor ligand supplied as a high-purity analytical/research standard and used as the precursor for fluorine-18 radioligand synthesis for PET imaging. It is provided as a 100 mg powder with documented solubility and storage recommendations for research use.
  • Selective 5-HT2A receptor ligand.
  • Suitable as an analytical and research standard.
  • Used to synthesize fluorine-18 radioligands for PET imaging.
  • High purity (99.93%) for experimental consistency.
  • Soluble in DMSO (requires warming/sonication; see handling notes).
  • Supplied as a 100 mg powder with recommended cold storage.
Catalog No. 50-002-28049