Learn More
Medchemexpress LLC Altanserin | 76330-71-7 | MFCD11045280 | 99.9% | 411.5 g/mol | C22H22FN3O2S | 50 MG

Supplier: Medchemexpress LLC HY11915650MG
Altanserin (CAS 76330-71-7) is a 5-HT2 receptor antagonist supplied as a high-purity analytical standard for research use, including neuroreceptor binding studies and synthesis of Fluorine-18 labeled tracer. The compound is provided as a 50 mg solid and is intended for laboratory research and analytical applications.
- High purity suitable for research and analytical use.
- Used to synthesize Fluorine-18 altanserin for PET imaging studies.
- Solid, white to light yellow appearance.
- Molecular weight 411.5 g/mol; formula C22H22FN3O2S.
- Soluble in DMSO at about 19.23 mg/mL.
- Stable when stored as powder at -20°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.