Learn More
JDM LAB SOLUTIONS LLC AlteCap™ Solo - External thread Micronic screw caps (96 SBS rack format)
Supplier:  JDM LAB SOLUTIONS LLC 404103
Encompass
Semi-automated single decapper. Electric capping & decapping. Compatible with 96 SBS rack format Micronic external thread screw caps - part numbers MP53825, MP53820, MP53801, MP53804, MP53800, MP53802, MP53809, MP53821, MP53803, MP53806, MP53823, MP53805, MP53822.Includes: Manual, stand, USB charging cable & cap driver.
Catalog No. 50-271-9977