Learn More
AdipoGen Altenusin (5 mg)
SDP
Supplier:  AdipoGen AGCN20143M005
Encompass_Preferred
Chemical. CAS: 31186-12-6. Formula: C15H14O6. MW: 290.3. Isolated from Penicillium sp. Antibiotic. Antifungal penicillide. Non-competitive, specific neutral sphingomyelinase (N-SMase) inhibitor. Strong pp60c-Src inhibitor. Inhibits cFMS receptor tyrosine kinase (CSF-1/m-CSF receptor tyrosine kinase) which is implicated in cancer and bone diseases. Myosin light chain kinase inhibitor. Exhibits anti-HIV-1 integrase activity. Cytotoxic activity against mouse lymphoma cells. Anti-protozoan, trypanothione reductase (TR) inhibitor. Phytotoxin.
Catalog No. 50-205-3523