Learn More
JDM LAB SOLUTIONS LLC AlteRead SBS rack 2D barcode camera reader (compatible with AcoustiX tubes)
Supplier:  JDM LAB SOLUTIONS LLC 252001
Encompass
Fast SBS 2D barcode camera rack reader. Decodes one full rack in < 1 seconds. SBS: 24, 48 & 96 racks. Decodes embedded 2D rack IDs. CryoCover reduces condensation for uninterrupted reading of cold racks. Includes: AlteRead software,Export data as XML, Excel, text (.csv or .txt Warranty: 5 years
Catalog No. 50-272-0020
Only null left
Add to Cart
Request a Quote