Learn More
Cayman Chemical AltretamIn 100mg
SDP
Supplier:  Cayman Chemical 23662100 MG
Encompass_Preferred
An anticancer agent; induces cytotoxicity in an ovarian cancer cell line at 10 μg/ml; inhibits GPX4 and induces accumulation of lipid-reactive oxygen species in U-2932 cells without depleting GSH levels at 0.5 mM; reduces tumor growth in an M5076 murine sarcoma model at 150 mg/kg
Catalog No. 50-299-5232