Learn More
Sorbent Technologies Inc Alumina, Basic, Active, 32-63um, 500g
SDP
Supplier:  Sorbent Technologies Inc 1556005
Encompass_Preferred
Alumina Basic, Active, is used for packing flash chromatography columns. It has a particle distribution of 32-63um and a quantity of 500g. The approximate pH is 10, Loss on Ignition (LOI) is < 1.7%, and specific surface area is about 200 m2/g.
Catalog No. 50-113-8754