Learn More
Perkin Elmer US LLC Autosampler Aluminum Sealed Sample Pans - 40 µL, Qty. 400
Supplier:  Perkin Elmer US LLC B0143021
Encompass_Preferred
These Autosampler Aluminum Sample Pans have a length of 10 mm, a depth of 2.1 mm, a volume of 40 µL, and an operating range of -170 to 600°C. They have a maximum pressure threshold of 3 bar. These pans require the Autosampler Aluminum Sample Covers (B0143003) and can be use with the Universal Crimper (B0139005). They are compatible with the DSC 6/7/4000/6000/8000/8500/Jade DSC/Diamond DSC/ PYRIS 1 DSC/PYRIS 6 DSC. Qty. 400 pans
Catalog No. 50-905-1975