Learn More
Accuflow Systems ALUM WEIGH DISH 12X4X2.7 250PK
Supplier:  Accuflow Systems MAWD1200
Encompass_Preferred
Eagle Thermoplastics - Alumimum Micro Weighing Dishes ,12W X4 L X2.7 H pack of 250
Catalog No. 50-109-4664