Learn More
Alkali Scientific AlumaFoil™ RollRight™, Dispenser +1 Roll of AlumaFoil Sheets, 500 Pre-Scored Sheets
SDP
Supplier:  Alkali Scientific SF16ML
Encompass
Self-standing, non-slip supports hold the pre-scored aluminum foil sheets film roll securely in place for quick and easy bench-top dispensement. The roll unwind tension can be easily adjusted to user preference. Continuous backing liner can be torn off or fed directly into a wastebasket for disposal. Certified DNase- & RNase- free. Pierceable with a pipet tip or robotic probe for direct sample recovery. 2.0mil aluminum foil removes easily without tearing. The conformable nature of foil allows for the visual verification of the seal’s security around each well. Not recommended for long-term cold storage.
Catalog No. 50-238-8687