Learn More
Sigma Aldrich Fine Chemicals Biosciences AlumaSealR II film non-ste
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A2350100EA
Encompass
A soft aluminum foil film designed for the sealing of PCR plates minimizing evaporation and vapor loss during thermal cycling. The film pierces very easily for direct sample recovery with a multichannel pipettor or robotic metal probes without risk of aerosols cross-contamination or significant gumming of adhesive. An extremely strong medical-grade adhesive provides a uniform complete seal which can withstand PCR thermal cycling. Recommended for temperatures from -80 C to 120 C measures 82.6 mm x 146.1 mm and offers sufficient sealing area for all PCR plates. A 36 mum non-permeable soft aluminum foil film eliminates the need for heat-sealing devices or mats during thermal cycling. The proprietary adhesive can withstand harsh chemicals solvents and reagents. Two end tabs for easy and accurate positioning and labeling.
Catalog No. 50-290-0887