Learn More
Advalue Technologies Alumina Combustion Boat
SDP
Supplier:  Advalue Technologies AL5003
Encompass
AL-5003 Alumina Combustion Boat - Capacity 3ml - Length 50mm - Width 12mm - Height 10mm
Catalog No. NC1048875