Learn More
Advalue Technologies Alumina Crucible Rectangular
SDP
Supplier:  Advalue Technologies AL3060
Encompass
AL-3060 Alumina Crucible Rectangular - Capacity 60ml - Length 82mm - Width 42mm - Height 23mm
Catalog No. NC1534857