Learn More
Electron Microscopy Sciences Alumina Powder Alpha, Type DX, 0.3 µm Particles 1 LB
SDP
Supplier:  Electron Microscopy Sciences 5036105
Encompass_Preferred
EMS's Alumina Powders are accurately controlled for all specs and particle size distribution, assuring you the best polishing results. Typical applications are precision optics, metallographic sections, electro-optical crystals, silicone wafers, ferrite components, gem stones, and acrylic lenses. Type DX Alumina Powder is specially treated to reduce the number of agglomerates.
Particles: 0.3 µm
Catalog No. 50-997-301