Learn More
Strem, An Ascensus Company CAS# 30745-55-2. 100g. Aluminum 2-ethylhexanoate, basic, min. 96%. MFCD00041940
SDP
Supplier:  Strem, An Ascensus Company 931326100G
Encompass
CAS# 30745-55-2. 100g. Aluminum 2-ethylhexanoate, basic, min. 96%. MFCD00041940. Molecular Weight: 330.41. Molecular Formula: AlOH[OOCCH(C2H5)C4H9]2. Color/form: white pwdr. Strem# 93-1326. http://www.strem.com/catalog/v/93-1326/
Catalog No. 50-901-14019