Learn More
Ethos Biosciences Aluminum Chloride 70PCT 16oz
Supplier:  Ethos Biosciences 352516
Encompass_Preferred
Aluminum Chloride solution is a clear to slightly hazy colorless to pale yellow aqueous solution This reagent is widely used as a coagulant and flocculant in water and watewater treatment a catalyst in organic synthesis pigments and paper products For laboratory and industrial use only
Catalog No. 50-008-19193