Learn More
Strem, An Ascensus Company CAS# 7446-70-0. 250g. Aluminum chloride, anhydrous, reagent, 99%. MFCD00003422
SDP
Supplier:  Strem, An Ascensus Company 931387250G
Encompass
CAS# 7446-70-0. 250g. Aluminum chloride, anhydrous, reagent, 99%. MFCD00003422. Molecular Weight: 133.34. Molecular Formula: AlCl3. Color/form: white to yellow pwdr. Strem# 93-1387. http://www.strem.com/catalog/v/93-1387/
Catalog No. 50-901-14065