Learn More
LabScientific Aluminum microtube cooling block: 96 wells (12 x 8 format) for 1.5 mL microtubes and vials
Supplier:  LabScientific 2079
Encompass
Aluminum Cooling Block (6 x 7.5"; 15 x 19 cm) 96 well format; 12 x 8 format for 1.5ml microtubes and vials.
Catalog No. NC9936821