Learn More
Medchemexpress LLC Aluminum diacetylsalicylate | 23413-80-1 | 99.0% | 10 G

Supplier: Medchemexpress LLC HYB217510G
Aspirin Aluminum is a novel intermolecular compound designed to inhibit gastrointestinal mucosal disorders induced by NSAIDs (non-steroidal anti-inflammatory agents). This compound is stable under recommended storage conditions. Keep container tightly sealed in a cool, well-ventilated area, avoiding inhalation and contact with eyes and skin.
- Inhibits gastrointestinal mucosal disorders
- Purity of 99.0%
- Appearance: White to off-white solid
- Powder storage: -20°C for 3 years, 4°C for 2 years
- In-solvent storage: -80°C for 6 months, -20°C for 1 month
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.