Learn More
Medchemexpress LLC Aluminum Glycinate | 13682-92-3 | C₂H₆AlNO₄ | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYA0271
Encompass_Preferred
Aluminum Glycinate is an organo-metallic compound that functions as an antacid. It can be used in research for indigestion, acid reflux, and ulcers. It releases aluminum ions in gastric juice, which react with phosphate in the intestine to form insoluble aluminum phosphate, thereby reducing phosphate absorption. It provides a rapid and lasting buffering effect, maintaining the stomach's pH value between 3.5-4.5, and protects the gastric mucosa from gastric acid damage. Its antacid activity is not affected by pepsin.
  • Organo-metallic compound
  • Functions as an antacid
  • Used in research for indigestion, acid reflux, and ulcers
  • Provides rapid and lasting buffering effect
  • Maintains stomach pH between 3.5-4.5
  • Protects gastric mucosa from acid damage
  • Antacid activity is not affected by pepsin
Catalog No. 50-003-58942