Learn More
Medchemexpress LLC Aluminum hydroxide (Al(OH)3) | 21645-51-2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYB15215G
Encompass_Preferred
Aluminum Hydroxide is an orally active, primary form of aluminum employed as an adjuvant. Research concerning aluminum hydroxide-based adjuvants encompasses their repository effect, pro-phagocytic effect, and activation of the pro-inflammatory NLRP3 pathway. Additionally, Aluminum Hydroxide serves as an adjuvant to offset the low intrinsic immunogenicity of subunit vaccines.
  • Orally active
  • Functions as an adjuvant
  • Investigated for its repository effect
  • Investigated for its pro-phagocytic effect
  • Activates the pro-inflammatory NLRP3 pathway
  • Compensates for low inherent immunogenicity in subunit vaccines
Catalog No. 50-004-11936