Learn More
Strem, An Ascensus Company CAS# 24304-00-5. 500g. Aluminum nitride, agglomerated powder, high purity. MFCD00003429
SDP
Supplier:  Strem, An Ascensus Company 931377500G
Encompass
CAS# 24304-00-5. 500g. Aluminum nitride, agglomerated powder, high purity. MFCD00003429. Molecular Weight: 40.99. Molecular Formula: AlN. Color/form: avg. 2.5-4.0 micron gray pwdr. (99% <50 microns) Strem# 93-1377. http://www.strem.com/catalog/v/93-1377/
Catalog No. 50-901-14058