Learn More
Strem, An Ascensus Company CAS# 24304-00-5. 500g. Aluminum nitride, deglomerated powder, high purity. MFCD00003429
SDP
Supplier:  Strem, An Ascensus Company 131378500G
Encompass
CAS# 24304-00-5. 500g. Aluminum nitride, deglomerated powder, high purity. MFCD00003429. Molecular Weight: 40.99. Molecular Formula: AlN. Color/form: 1.0-15.0 micron gray pwdr. Strem# 13-1378. http://www.strem.com/catalog/v/13-1378/
Catalog No. 50-901-09546