Learn More
Electron Microscopy Sciences Aluminum Oxide 90 Basic 500 GR
SDP
Supplier:  Electron Microscopy Sciences 10400
Encompass_Preferred
CAS #1344-28-1 Al₂O₃ F.W. 101.96
Aluminum Oxide 90 Basic (63-200 Micron) for use with Technovit 9100.
Catalog No. 50-189-8709