Learn More
Strem, An Ascensus Company CAS# 1344-28-1. 500g. Aluminum oxide, activated, basic, gamma, 96%. MFCD00003424
SDP
Supplier:  Strem, An Ascensus Company 931334500G
Encompass
CAS# 1344-28-1. 500g. Aluminum oxide, activated, basic, gamma, 96%. MFCD00003424. Molecular Weight: 101.96. Molecular Formula: Al2O3. Color/form: -60 mesh white pwdr. Strem# 93-1334. http://www.strem.com/catalog/v/93-1334/
Catalog No. 50-901-14025