Learn More
Sigma Aldrich Fine Chemicals Biosciences Aluminum oxide activated a5KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1999665KG
Encompass
The mechanism of formation of protective scales of Al2O3 (aluminum oxide) on surfaces of binary M-Al (M fe Ni or Co) alloys at high temperatures has been studied. Activated aluminum oxides having different acidities and water concentrations (having Brockmann activity I-V) can easily detect various types of water (namely free water pore water and bound water) on the aluminum oxide present in soils.
Catalog No. 50-289-9034