Learn More
Medchemexpress LLC Aluminum phthalocyanine chloride | 14154-42-8 | 100 MG
Supplier: Medchemexpress LLC HY129959100MG
Aluminum phthalocyanine chloride is a photosensitizer that effectively inhibits the parasite *Leishmania amazonensis*, the causative agent of cutaneous leishmaniasis, through light-mediated cytolysis. It causes parasite morphology and cytolysis of isolated amastigotes and lyses parasites within infected J774 macrophages.
- Effectively inhibits *Leishmania amazonensis*
- Causes parasite morphology and cytolysis of isolated amastigotes
- Lyses parasites within infected J774 macrophages
- Can be used to investigate leishmaniasis
- Appears as a solid
- Color is blue to dark blue