Learn More
Medchemexpress LLC Aluminum phthalocyanine chloride | 14154-42-8 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY129959
Encompass_Preferred
Aluminum phthalocyanine chloride is a photosensitizer that effectively inhibits the parasite Leishmania amazonensis, the causative agent of cutaneous leishmaniasis, through light-mediated cytolysis. It induces morphological changes and cytolysis in isolated amastigotes and can lyse parasites within infected J774 macrophages.
  • Functions as a photosensitizer
  • Inhibits Leishmania amazonensis
  • Induces cytolysis in isolated amastigotes
  • Lyses parasites within infected J774 macrophages
  • Suitable for leishmaniasis research
  • For research use only
Catalog No. 50-003-42750