Learn More
Medchemexpress LLC Aluminum phthalocyanine chloride | 14154-42-8 | 50 MG
Supplier: Medchemexpress LLC HY12995950MG
Aluminum phthalocyanine chloride is a photosensitizer that effectively inhibits the parasite *Leishmania amazonensis*, the causative agent of cutaneous leishmaniasis. It achieves this through light-mediated cytolysis, inducing changes in parasite morphology and cytolysis of isolated amastigotes. This compound can also lyse parasites found within infected J774 macrophages, making it a valuable tool for research into leishmaniasis.
- Effectively inhibits *Leishmania amazonensis*
- Induces changes in parasite morphology and cytolysis
- Useful for research into leishmaniasis
- Requires higher photosensitizer concentrations and light intensities to induce lysis of mammalian cells