Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aluminum potassium sulfate | 10043-67-1 | MFCD00003428 | 99.6% | 258.21 g/mol | KAl(SO4)2 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW13430110G

Encompass_Preferred

Aluminum potassium sulfate (potassium alum) is an inorganic reagent used as a mordant, water purification agent, and antibacterial compound in laboratory and environmental research. It is supplied as a solid reagent with a high assay suitable for staining, microbiology, and water treatment studies.

  • High purity of 99.6%.
  • Chemical formula KAl(SO4)2.
  • CAS number 10043-67-1.
  • Functions as a mordant in staining applications.
  • Useful for water purification and antibacterial research.
  • Available in small laboratory pack sizes for research use.

Catalog No. 50-002-83302


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.