Learn More
Medchemexpress LLC Aluminum potassium sulfate | 10043-67-1 | MFCD00003428 | 99.6% | 258.21 g/mol | KAl(SO4)2 | 25 G
SDP
Supplier:  Medchemexpress LLC HYW13430125G
Encompass_Preferred
Aluminum potassium sulfate (potassium alum) is an inorganic salt supplied as a white crystalline solid for laboratory and industrial use. It functions as a flocculant and antibacterial agent and is commonly used in water purification, analytical methods, and reagent preparation. The material is offered at laboratory-grade purity for research workflows.
  • High purity: 99.6%.
  • White crystalline solid, water-soluble.
  • Used as a flocculant for water purification.
  • Provides antibacterial properties in applications.
  • Suitable for analytical and preparative laboratory use.
  • Available in laboratory pack sizes for research workflows.
Catalog No. 50-002-83400