Learn More
Medchemexpress LLC Aluminum potassium sulfate | 10043-67-1 | MFCD00003428 | 99.6% | 258.21 | KAl(SO4)2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1343015G
Encompass_Preferred
Aluminum potassium sulfate is an inorganic salt commonly used as a laboratory reagent for water purification, antimicrobial applications, and various biochemical protocols. It is supplied as a white to off-white crystalline solid with high purity suitable for research use.
  • High purity: 99.6%.
  • White to off-white solid, crystalline form.
  • Soluble in water with ultrasonic warming and heating to aid dissolution.
  • Stable when stored dry at room temperature; keep cool.
  • Useful for water purification, antimicrobial assays, and reagent preparation.
  • Available in small research pack sizes (5 G).
Catalog No. 50-002-83385