Learn More
Medchemexpress LLC Aluminum potassium sulfate dodecahydrate | 7784-24-9 | 98.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB2241A10G
Encompass_Preferred
Potassiumalum, for cell culture is an egg white adjuvant that functions as a mordant, aiding dyes in binding to tissue components. It enhances the staining effect and stability during the staining process. This product has been observed to induce allergic reactions in mice and is suitable for research applications, specifically in bacterial and tissue staining.
  • Functions as an egg white adjuvant
  • Aids dyes in binding to tissue components
  • Enhances staining effect and stability during staining
  • Suitable for research applications
  • Used in bacterial and tissue staining
  • Induces allergic reactions in mice
Catalog No. 50-004-10114