Learn More
Electron Microscopy Sciences Aluminum Tissue Mounting Stub
SDP
Supplier:  Electron Microscopy Sciences 7014510
Encompass_Preferred
Machined from aluminum rods with one end tapered to 4.8 mm in diameter. The specimen can be oriented and mounted to the tip with glue and it is ready for sectioning. The body size is "00" and it measures 11 mm long.
Catalog No. 50-293-60